Learn More
Cayman Chemical PhosphodISTRase 4 InhIbItr 1mg
SDP
Catalog No. 502982099
Encompass_Preferred
An inhibitor of PDE4 (IC50 = 0.10 μM for the human recombinant enzyme)
Catalog No. 50-298-2099 Supplier Cayman Chemical Supplier No. 211911 MG
An inhibitor of PDE4 (IC50 = 0.10 μM for the human recombinant enzyme)