Learn More
Cayman Chemical PhosphodISTRase 4 InhIbItr 5mg
SDP
Catalog No. 502983117
Encompass_Preferred
An inhibitor of PDE4 (IC50 = 0.10 μM for the human recombinant enzyme)
Catalog No. 50-298-3117 Supplier Cayman Chemical Supplier No. 211915 MG
Add to Cart
Add to Cart
An inhibitor of PDE4 (IC50 = 0.10 μM for the human recombinant enzyme)