Learn More
Bachem Phosphoenolpyruvate potassium salt
SDP
Supplier:  Bachem 4014027.0005
Encompass_Preferred
Phosphoenolpyruvate potassium salt, 4265-07-0, C3H4KO6P, Mr 206.13, 5g. Potassium phosphoenolpyruvate, PEP-K, 2-Phosphonooxy-acrylic acid potassium salt. Substrate for many kinases such as pyruvate kinase and phosphoenolpyruvate carboxykinase.
Catalog No. 50-260-633