Learn More
Sigma Aldrich Fine Chemicals Biosciences Phosphoenolpyruvc acd mo250MG
Catalog No. 502902596
Encompass
Phospho(enol)pyruvic acid (PEP) acts as an inhibitor of hexokinase phosphoglucose isomerase phosphofructokinase and aldolase. It is a bifunctional carbohydrate which exhibits antioxidant property. PEP may be a potential organ preservation agent in clinical transplantation.
Catalog No. 50-290-2596 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P7127250MG
Phospho(enol)pyruvic acid (PEP) acts as an inhibitor of hexokinase phosphoglucose isomerase phosphofructokinase and aldolase. It is a bifunctional carbohydrate which exhibits antioxidant property. PEP may be a potential organ preservation agent in clinical transplantation.