Learn More
Medchemexpress LLC HY-K0221 60 mL Medchemexpress, Affinity Chromatography Column CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYK022160 ML5 PCS
Encompass_Preferred
Medchemexpress, HY-W011704A 10mg Phosphoenolpyruvic acid (tricyclohexylammoniu​m salt) CAS:35556-70-8 Phosphoenolpyruvic acid tricyclohexylammoniu​m salt is a glycolysis metabolite with a high-energy phosphate group, penetrates the cell membrane and exhibits cytoprotective and anti-oxidative activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0210