Learn More
Medchemexpress LLC Phosphoenolpyruvic acid cyclohexylammonium salt | 10526-80-4 | 98.0% | 267.22 | 1 ML
SDP
Catalog No. 5000452475
Encompass_Preferred
Phosphoenolpyruvic acid cyclohexylammonium salt is a glycolysis metabolite featuring a high-energy phosphate group. It participates in glycolysis and gluconeogenesis, serving as an energy source to generate adenosine triphosphate (ATP) under energy-limited conditions. Additionally, this salt exhibits cytoprotective and anti-oxidative qualities.
  • Involved in glycolysis and gluconeogenesis.
  • Functions as an energy source to produce ATP.
  • Possesses cytoprotective properties.
  • Demonstrates anti-oxidative properties.
  • Significantly attenuates the decrease in cell viability induced by hydrogen peroxide in HeLa cells in a dose-dependent manner.
  • Significantly reduces hydrogen peroxide-stimulated increase in intracellular reactive oxygen species.
  • Demonstrates scavenging potential against hydroxyl radicals.
  • Attenuates cell injury induced by 2-deoxy-D-glucose.
Catalog No. 50-004-52475 Supplier Medchemexpress LLC Supplier No. HYW01170410MM/1ML
Phosphoenolpyruvic acid cyclohexylammonium salt is a glycolysis metabolite featuring a high-energy phosphate group. It participates in glycolysis and gluconeogenesis, serving as an energy source to generate adenosine triphosphate (ATP) under energy-limited conditions. Additionally, this salt exhibits cytoprotective and anti-oxidative qualities.
  • Involved in glycolysis and gluconeogenesis.
  • Functions as an energy source to produce ATP.
  • Possesses cytoprotective properties.
  • Demonstrates anti-oxidative properties.
  • Significantly attenuates the decrease in cell viability induced by hydrogen peroxide in HeLa cells in a dose-dependent manner.
  • Significantly reduces hydrogen peroxide-stimulated increase in intracellular reactive oxygen species.
  • Demonstrates scavenging potential against hydroxyl radicals.
  • Attenuates cell injury induced by 2-deoxy-D-glucose.