Learn More
Medchemexpress LLC Phosphoenolpyruvic acid cyclohexylammonium salt | 10526-80-4 | 98.0% | 267.22 | 100 MG
SDP
Catalog No. 5000453676
Encompass_Preferred
Phosphoenolpyruvic acid cyclohexylammonium salt is a glycolysis metabolite with a high-energy phosphate group. It is involved in glycolysis and gluconeogenesis, serving as an energy source to produce ATP under energy-limited conditions. This compound also exhibits cytoprotective and anti-oxidative properties.
  • Reduces decrease in cell viability induced by hydrogen peroxide.
  • Reduces hydrogen peroxide stimulated increase in intracellular reactive oxygen species.
  • Demonstrates scavenging potential against hydroxyl radicals.
  • Inhibits decrease in cellular ATP content induced by hydrogen peroxide (at 10 mM).
  • Attenuates cell injury induced by 2-Deoxy-D-glucose.
Catalog No. 50-004-53676 Supplier Medchemexpress LLC Supplier No. HYW011704100MG
Phosphoenolpyruvic acid cyclohexylammonium salt is a glycolysis metabolite with a high-energy phosphate group. It is involved in glycolysis and gluconeogenesis, serving as an energy source to produce ATP under energy-limited conditions. This compound also exhibits cytoprotective and anti-oxidative properties.
  • Reduces decrease in cell viability induced by hydrogen peroxide.
  • Reduces hydrogen peroxide stimulated increase in intracellular reactive oxygen species.
  • Demonstrates scavenging potential against hydroxyl radicals.
  • Inhibits decrease in cellular ATP content induced by hydrogen peroxide (at 10 mM).
  • Attenuates cell injury induced by 2-Deoxy-D-glucose.