Learn More
Medchemexpress LLC Phosphoenolpyruvic acid cyclohexylammonium salt | 10526-80-4 | 98.0% | 267.22 | 500 MG
SDP
Catalog No. 5000452868
Encompass_Preferred
Phosphoenolpyruvic acid (Phosphoenolpyruvate) cyclohexylammonium salt is a glycolysis metabolite with a high-energy phosphate group. It is involved in glycolysis and gluconeogenesis, and used as an energy source to produce ATP under energy-limited conditions. This compound also exhibits cytoprotective and anti-oxidative properties. It is for research use only.
  • Glycolysis metabolite with a high-energy phosphate group
  • Involved in glycolysis and gluconeogenesis
  • Used as an energy source to produce ATP under energy-limited conditions
  • Exhibits cytoprotective and anti-oxidative properties
  • Significantly reduces the decrease in cell viability induced by hydrogen peroxide
  • Reduces H2O2-stimulated increase in intracellular reactive oxygen species
  • Demonstrates scavenging potential against hydroxyl radicals
  • Attenuates cell injury induced by 2-Deoxy-D-glucose
Catalog No. 50-004-52868 Supplier Medchemexpress LLC Supplier No. HYW011704500MG
Phosphoenolpyruvic acid (Phosphoenolpyruvate) cyclohexylammonium salt is a glycolysis metabolite with a high-energy phosphate group. It is involved in glycolysis and gluconeogenesis, and used as an energy source to produce ATP under energy-limited conditions. This compound also exhibits cytoprotective and anti-oxidative properties. It is for research use only.
  • Glycolysis metabolite with a high-energy phosphate group
  • Involved in glycolysis and gluconeogenesis
  • Used as an energy source to produce ATP under energy-limited conditions
  • Exhibits cytoprotective and anti-oxidative properties
  • Significantly reduces the decrease in cell viability induced by hydrogen peroxide
  • Reduces H2O2-stimulated increase in intracellular reactive oxygen species
  • Demonstrates scavenging potential against hydroxyl radicals
  • Attenuates cell injury induced by 2-Deoxy-D-glucose