Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoenolpyruvic acid monopotassium salt, 99%, Thermo Scientific™

Catalog No. AC229100010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC229100010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC229100010 Supplier Thermo Scientific Chemicals Supplier No. 229100010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4265-07-0
Molecular Formula C3H4KO6P
Molecular Weight (g/mol) 206.131
InChI Key SOSDSEAIODNVPX-UHFFFAOYSA-M
Synonym potassium 1-carboxyvinyl hydrogenphosphate, potassium hydrogen 2-phosphonatooxy acrylate, 2-propenoic acid, 2-phosphonooxy-, monopotassium salt, potassium 1-carboxyvinyl hydrogen phosphate, phospho enol pyruvic acid monopotassium salt, phosphoenolpyruvic acid monopotassium salt, 2-propenoic acid, 2-phosphonooxy-, potassium salt 1:1, potassium 2-hydrogen phosphonooxy prop-2-enoic acid, phosphoenolpyruvate potassium salt, potassium 2-hydrogen phosphonatooxy prop-2-enoic acid
PubChem CID 23678879
IUPAC Name potassium;1-carboxyethenyl hydrogen phosphate
SMILES C=C(C(=O)O)OP(=O)(O)[O-].[K+]

Specifications

Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula KOP(O)(OH)OC(=CH2)CO2H
Quantity 1 g
Beilstein 03, IV, 977
Formula Weight 206.13
Percent Purity ≥98.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material Phosphoenolpyruvic acid monopotassium salt, 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.