Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoenolpyruvic acid, monopotassium salt, MP Biomedicals™

Catalog No. ICN19570791 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
0.1 g
250 mg
1 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19570791 1 g
ICN19570783 25 mg
ICN19570725 250 mg
ICN19570780 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19570791 Supplier MP Biomedicals, Inc Supplier No. 0219570791
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4265-07-0
Molecular Formula C3H4KO6P
Molecular Weight (g/mol) 206.131
MDL Number MFCD00044476
InChI Key SOSDSEAIODNVPX-UHFFFAOYSA-M
Synonym potassium 1-carboxyvinyl hydrogenphosphate, potassium hydrogen 2-phosphonatooxy acrylate, 2-propenoic acid, 2-phosphonooxy-, monopotassium salt, potassium 1-carboxyvinyl hydrogen phosphate, phospho enol pyruvic acid monopotassium salt, phosphoenolpyruvic acid monopotassium salt, 2-propenoic acid, 2-phosphonooxy-, potassium salt 1:1, potassium 2-hydrogen phosphonooxy prop-2-enoic acid, phosphoenolpyruvate potassium salt, potassium 2-hydrogen phosphonatooxy prop-2-enoic acid
PubChem CID 23678879
IUPAC Name potassium;1-carboxyethenyl hydrogen phosphate
SMILES C=C(C(=O)O)OP(=O)(O)[O-].[K+]

Specifications

Color White
Quantity 1 g
Formula Weight 206.1
Physical Form Crystalline Powder
Chemical Name or Material Phosphoenolpyruvic acid, monopotassium salt
Recommended Storage Store at -0°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.