Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoenolpyruvic Acid Monopotassium Salt, MP Biomedicals™

Catalog No. MP219570794 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219570794 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219570794 Supplier MP Biomedicals, Inc Supplier No. 0219570794
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Functions as an important metabolic intermediate in animals, plants, and microorganisms. Phosphoenolpyruvic Acid Monopotassium Salt, MP Biomedicals is a phosphoric ester of the enol form of pyruvic acid. PEP is a high-energy compound involved in glycolysi

Phospho(enol)pyruvic acid (PEP) is involved in glycolysis and gluconeogeneis. In glycolysis, PEP is metabolized by Pyruvate Kinase to yield pyruvate. In plants, PEP is involved in the formation of aromatic amino acids as well as in the carbon fixation pathway.

PEP is used for regeneration of ATP from ADP in enzyme-catalyzed syntheses. PEP protects against the cell injury induced by oxidative stress and against the dysfunction in carbohydrate metabolism.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4265-07-0
Molecular Formula C3H4KO6P
Molecular Weight (g/mol) 206.131
InChI Key SOSDSEAIODNVPX-UHFFFAOYSA-M
Synonym potassium 1-carboxyvinyl hydrogenphosphate, potassium hydrogen 2-phosphonatooxy acrylate, 2-propenoic acid, 2-phosphonooxy-, monopotassium salt, potassium 1-carboxyvinyl hydrogen phosphate, phospho enol pyruvic acid monopotassium salt, phosphoenolpyruvic acid monopotassium salt, 2-propenoic acid, 2-phosphonooxy-, potassium salt 1:1, potassium 2-hydrogen phosphonooxy prop-2-enoic acid, phosphoenolpyruvate potassium salt, potassium 2-hydrogen phosphonatooxy prop-2-enoic acid
PubChem CID 23678879
IUPAC Name potassium;1-carboxyethenyl hydrogen phosphate
SMILES C=C(C(=O)O)OP(=O)(O)[O-].[K+]

Specifications

Melting Point 175°C (literature)(decomposition)
Quantity 5 g
Solubility Information Soluble in water (100mg/mL)
Formula Weight 206.1g/mol
Physical Form Crystalline Powder
Recommended Storage Store at 0°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.