Learn More
Medchemexpress LLC Phosphoenolpyruvic acid potassium-13C2 | 201996-39-6 | 208.12 | 10 MG
SDP
Catalog No. 5000453146
Encompass_Preferred
Phosphoenolpyruvic acid (potassium)-13C2 is the 13C labeled Phosphoenolpyruvic acid potassium (HY-W008807). Phosphoenolpyruvic acid (Phosphoenolpyruvate) potassium is a glycolysis metabolite with a high-energy phosphate group. It is involved in glycolysis and gluconeogenesis, and used as an energy source to produce ATP under energy-limited conditions. This compound also exhibits cytoprotective and anti-oxidative properties. It is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-53146 Supplier Medchemexpress LLC Supplier No. HYW008807S10MG
Phosphoenolpyruvic acid (potassium)-13C2 is the 13C labeled Phosphoenolpyruvic acid potassium (HY-W008807). Phosphoenolpyruvic acid (Phosphoenolpyruvate) potassium is a glycolysis metabolite with a high-energy phosphate group. It is involved in glycolysis and gluconeogenesis, and used as an energy source to produce ATP under energy-limited conditions. This compound also exhibits cytoprotective and anti-oxidative properties. It is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.