Learn More
Medchemexpress LLC Phosphoenolpyruvic acid potassium-13C2 | 201996-39-6 | 97.0% | 208.12 | 1 MG
SDP
Catalog No. 5000366620
Encompass_Preferred
Phosphoenolpyruvic acid (potassium)-13C2 is the 13C labeled version of Phosphoenolpyruvic acid potassium. This compound is a glycolysis metabolite that contains a high-energy phosphate group. It participates in both glycolysis and gluconeogenesis, serving as an energy source to generate ATP under conditions of limited energy. Additionally, Phosphoenolpyruvic acid potassium has demonstrated cytoprotective and anti-oxidative properties.
  • 13C labeled compound, useful as a tracer.
  • Can be utilized as an internal standard for quantitative analysis using methods like NMR, GC-MS, or LC-MS.
  • Involved in glycolysis and gluconeogenesis.
  • Acts as an energy source for ATP production in energy-limited conditions.
  • Exhibits cytoprotective properties.
  • Shows anti-oxidative properties.
Catalog No. 50-003-66620 Supplier Medchemexpress LLC Supplier No. HYW008807S1MG
Phosphoenolpyruvic acid (potassium)-13C2 is the 13C labeled version of Phosphoenolpyruvic acid potassium. This compound is a glycolysis metabolite that contains a high-energy phosphate group. It participates in both glycolysis and gluconeogenesis, serving as an energy source to generate ATP under conditions of limited energy. Additionally, Phosphoenolpyruvic acid potassium has demonstrated cytoprotective and anti-oxidative properties.
  • 13C labeled compound, useful as a tracer.
  • Can be utilized as an internal standard for quantitative analysis using methods like NMR, GC-MS, or LC-MS.
  • Involved in glycolysis and gluconeogenesis.
  • Acts as an energy source for ATP production in energy-limited conditions.
  • Exhibits cytoprotective properties.
  • Shows anti-oxidative properties.