Learn More
Medchemexpress LLC Phosphoenolpyruvic acid potassium-13C2 | 201996-39-6 | 97.0% | 208.12 | 5 MG
SDP
Catalog No. 5000451949
Encompass_Preferred
Phosphoenolpyruvic acid (potassium)-13C2 is the 13C labeled Phosphoenolpyruvic acid potassium. Phosphoenolpyruvic acid (Phosphoenolpyruvate) potassium is a glycolysis metabolite with a high-energy phosphate group. Phosphoenolpyruvic acid potassium is involved in glycolysis and gluconeogenesis, and used as energy source to produce ATP, under the energy-limited conditions. Phosphoenolpyruvic acid potassium also exhibits cytoprotective and anti-oxidative properties.
  • Glycolysis metabolite with a high-energy phosphate group.
  • Involved in glycolysis and gluconeogenesis.
  • Used as energy source to produce ATP under energy-limited conditions.
  • Exhibits cytoprotective and anti-oxidative properties.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-51949 Supplier Medchemexpress LLC Supplier No. HYW008807S5MG
Phosphoenolpyruvic acid (potassium)-13C2 is the 13C labeled Phosphoenolpyruvic acid potassium. Phosphoenolpyruvic acid (Phosphoenolpyruvate) potassium is a glycolysis metabolite with a high-energy phosphate group. Phosphoenolpyruvic acid potassium is involved in glycolysis and gluconeogenesis, and used as energy source to produce ATP, under the energy-limited conditions. Phosphoenolpyruvic acid potassium also exhibits cytoprotective and anti-oxidative properties.
  • Glycolysis metabolite with a high-energy phosphate group.
  • Involved in glycolysis and gluconeogenesis.
  • Used as energy source to produce ATP under energy-limited conditions.
  • Exhibits cytoprotective and anti-oxidative properties.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.