Learn More
Medchemexpress LLC Phosphoenolpyruvic acid potassium | 4265-07-0 | 99.86% | 206.13 | 5 G
SDP
Catalog No. 5000451200
Encompass_Preferred
Phosphoenolpyruvic acid potassium is a glycolysis metabolite featuring a high-energy phosphate group. It plays a crucial role in glycolysis and gluconeogenesis, serving as an energy source to produce ATP under energy-limited conditions.
  • Involved in glycolysis and gluconeogenesis.
  • Functions as an energy source to produce ATP.
  • Exhibits cytoprotective properties.
  • Shows anti-oxidative properties.
Catalog No. 50-004-51200 Supplier Medchemexpress LLC Supplier No. HYW0088075G
Phosphoenolpyruvic acid potassium is a glycolysis metabolite featuring a high-energy phosphate group. It plays a crucial role in glycolysis and gluconeogenesis, serving as an energy source to produce ATP under energy-limited conditions.
  • Involved in glycolysis and gluconeogenesis.
  • Functions as an energy source to produce ATP.
  • Exhibits cytoprotective properties.
  • Shows anti-oxidative properties.