Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phosphoenolpyruvic acid potassium | 4265-07-0 | 99.90% | 206.13 | 500 MG
SDP

Catalog No. 5000452163
Encompass_Preferred

Phosphoenolpyruvic acid potassium is a glycolysis metabolite with a high-energy phosphate group. It is involved in glycolysis and gluconeogenesis, and used as an energy source to produce ATP under energy-limited conditions. It also exhibits cytoprotective and anti-oxidative properties.

  • Glycolysis metabolite
  • Exhibits cytoprotective and anti-oxidative properties
  • Involved in glycolysis and gluconeogenesis
  • Used as energy source to produce ATP
  • Appearance: solid, white to off-white
  • Initial source: endogenous metabolite
  • Shipping: room temperature in continental US; may vary elsewhere.
  • Storage: 4°C, sealed storage, away from moisture.

Catalog No. 50-004-52163 Supplier Medchemexpress LLC Supplier No. HYW008807500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phosphoenolpyruvic acid potassium is a glycolysis metabolite with a high-energy phosphate group. It is involved in glycolysis and gluconeogenesis, and used as an energy source to produce ATP under energy-limited conditions. It also exhibits cytoprotective and anti-oxidative properties.

  • Glycolysis metabolite
  • Exhibits cytoprotective and anti-oxidative properties
  • Involved in glycolysis and gluconeogenesis
  • Used as energy source to produce ATP
  • Appearance: solid, white to off-white
  • Initial source: endogenous metabolite
  • Shipping: room temperature in continental US; may vary elsewhere.
  • Storage: 4°C, sealed storage, away from moisture.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.