Learn More
Medchemexpress LLC Phosphoenolpyruvic acid potassium | 4265-07-0 | MFCD00044476 | 99.9% | 206.13 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW008807R
Encompass_Preferred
Phosphoenolpyruvic acid potassium is a glycolysis metabolite with a high-energy phosphate group. It is involved in glycolysis and gluconeogenesis, and used as an energy source to produce ATP under energy-limited conditions. It also exhibits cytoprotective and anti-oxidative properties.
  • Attenuates decrease in cell viability induced by hydrogen peroxide in HeLa cells.
  • Reduces hydrogen peroxide-stimulated increase in intracellular reactive oxygen species.
  • Demonstrates scavenging potential against hydroxyl radicals.
  • Slightly inhibits decrease in cellular ATP content induced by hydrogen peroxide.
  • Attenuates cell injury.
Catalog No. 50-004-54037