Learn More
Medchemexpress LLC Phosphoenolpyruvic acid potassium (Standard) | 4265-07-0 | 99.0% | 206.13 | 10 MG
SDP
Catalog No. 5000453668
Encompass_Preferred
Phosphoenolpyruvic acid potassium (Standard) is an analytical standard used for research and analytical applications. It is a glycolytic metabolite with a high-energy phosphate group, involved in glycolysis and gluconeogenesis, serving as an energy source for ATP production under energy-limited conditions. It also exhibits cytoprotective and antioxidant properties.
  • Used as an analytical standard for content determination
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Suitable for liquid chromatography, gas chromatography, and mass spectrometry
Catalog No. 50-004-53668 Supplier Medchemexpress LLC Supplier No. HYW008807R10MG
Phosphoenolpyruvic acid potassium (Standard) is an analytical standard used for research and analytical applications. It is a glycolytic metabolite with a high-energy phosphate group, involved in glycolysis and gluconeogenesis, serving as an energy source for ATP production under energy-limited conditions. It also exhibits cytoprotective and antioxidant properties.
  • Used as an analytical standard for content determination
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Suitable for liquid chromatography, gas chromatography, and mass spectrometry