Learn More
Medchemexpress LLC Phosphoenolpyruvic acid tricyclohexylammonium salt | 35556-70-8 | 99.66% | 465.56 | 1 ML
SDP
Catalog No. 5000453902
Encompass_Preferred
Phosphoenolpyruvic acid (Phosphoenolpyruvate) tricyclohexylammonium salt is a glycolysis metabolite with a high-energy phosphate group. It is involved in glycolysis and gluconeogenesis, and used as an energy source to produce ATP, under energy-limited conditions. This compound also exhibits cytoprotective and anti-oxidative properties. It is for research use only.
  • Glycolysis metabolite
  • High-energy phosphate group
  • Involved in glycolysis and gluconeogenesis
  • Used as an energy source to produce ATP
  • Exhibits cytoprotective properties
  • Exhibits anti-oxidative properties
  • For research use only
Catalog No. 50-004-53902 Supplier Medchemexpress LLC Supplier No. HYW011704A10MM/1ML
Phosphoenolpyruvic acid (Phosphoenolpyruvate) tricyclohexylammonium salt is a glycolysis metabolite with a high-energy phosphate group. It is involved in glycolysis and gluconeogenesis, and used as an energy source to produce ATP, under energy-limited conditions. This compound also exhibits cytoprotective and anti-oxidative properties. It is for research use only.
  • Glycolysis metabolite
  • High-energy phosphate group
  • Involved in glycolysis and gluconeogenesis
  • Used as an energy source to produce ATP
  • Exhibits cytoprotective properties
  • Exhibits anti-oxidative properties
  • For research use only