Learn More
Medchemexpress LLC Phosphoenolpyruvic acid tricyclohexylammonium salt | 35556-70-8 | 99.7% | 465.56 | 5 MG
SDP
Catalog No. 5000452566
Encompass_Preferred
Phosphoenolpyruvic acid tricyclohexylammonium salt is a glycolysis metabolite featuring a high-energy phosphate group. It plays a role in both glycolysis and gluconeogenesis, serving as an energy source for ATP production under energy-limited conditions. Additionally, this compound demonstrates cytoprotective and anti-oxidative properties.
  • Glycolysis metabolite.
  • Contains a high-energy phosphate group.
  • Involved in glycolysis and gluconeogenesis.
  • Provides energy for ATP synthesis.
  • Exhibits cytoprotective properties.
  • Offers anti-oxidative benefits.
Catalog No. 50-004-52566 Supplier Medchemexpress LLC Supplier No. HYW011704A5MG
Phosphoenolpyruvic acid tricyclohexylammonium salt is a glycolysis metabolite featuring a high-energy phosphate group. It plays a role in both glycolysis and gluconeogenesis, serving as an energy source for ATP production under energy-limited conditions. Additionally, this compound demonstrates cytoprotective and anti-oxidative properties.
  • Glycolysis metabolite.
  • Contains a high-energy phosphate group.
  • Involved in glycolysis and gluconeogenesis.
  • Provides energy for ATP synthesis.
  • Exhibits cytoprotective properties.
  • Offers anti-oxidative benefits.