Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phosphoenolpyruvic acid (tricyclohexylammonium salt) | 35556-70-8 | 99.7% | 465.56 | 5G
SDP

Catalog No. 5000452429
Encompass_Preferred

Phosphoenolpyruvic acid tricyclohexylammonium salt is a glycolysis metabolite with a high-energy phosphate group. It is involved in glycolysis and gluconeogenesis, and used as an energy source to produce ATP under energy-limited conditions. This compound also exhibits cytoprotective and anti-oxidative properties. It is for research use only.

  • Glycolysis metabolite with a high-energy phosphate group
  • Involved in glycolysis and gluconeogenesis
  • Used as an energy source to produce ATP under energy-limited conditions
  • Exhibits cytoprotective and anti-oxidative properties
  • Appearance: Solid, White to off-white
  • Initial source: Endogenous metabolite
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: -20°C, sealed storage, away from moisture

Catalog No. 50-004-52429 Supplier Medchemexpress LLC Supplier No. HYW0117045G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phosphoenolpyruvic acid tricyclohexylammonium salt is a glycolysis metabolite with a high-energy phosphate group. It is involved in glycolysis and gluconeogenesis, and used as an energy source to produce ATP under energy-limited conditions. This compound also exhibits cytoprotective and anti-oxidative properties. It is for research use only.

  • Glycolysis metabolite with a high-energy phosphate group
  • Involved in glycolysis and gluconeogenesis
  • Used as an energy source to produce ATP under energy-limited conditions
  • Exhibits cytoprotective and anti-oxidative properties
  • Appearance: Solid, White to off-white
  • Initial source: Endogenous metabolite
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: -20°C, sealed storage, away from moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.