Learn More
Medchemexpress LLC Phosphoglucose isomerase, rabbit muscle (GPI) | 9001-41-6 | ≥200 U/mg protein | 1 KU
SDP
Catalog No. 5000398708
Encompass_Preferred
Phosphoglucose Isomerase, Rabbit muscle (GPI), also known as phosphoglucoisomerase (PGI) or phosphohexose isomerase (PHI), is frequently utilized in biochemical studies. This enzyme is a highly conserved glycolytic and dimeric enzyme that catalyzes the reversible isomerization of glucose-6-phosphate to fructose-6-phosphate.
  • Utilized in biochemical studies
  • Highly conserved glycolytic and dimeric enzyme
  • Catalyzes reversible isomerization of glucose-6-phosphate to fructose-6-phosphate
Catalog No. 50-003-98708 Supplier Medchemexpress LLC Supplier No. HYP29181KU
Phosphoglucose Isomerase, Rabbit muscle (GPI), also known as phosphoglucoisomerase (PGI) or phosphohexose isomerase (PHI), is frequently utilized in biochemical studies. This enzyme is a highly conserved glycolytic and dimeric enzyme that catalyzes the reversible isomerization of glucose-6-phosphate to fructose-6-phosphate.
  • Utilized in biochemical studies
  • Highly conserved glycolytic and dimeric enzyme
  • Catalyzes reversible isomerization of glucose-6-phosphate to fructose-6-phosphate