Learn More
Biomatik Corporation Phospho-Lat (Y191) Recombinant Antibody
Supplier:  Biomatik Corporation RAC07013
Encompass_Preferred
Phospho-Lat (Y191) Recombinant Antibody is a Human (Homo sapiens) Recombinant Antibody to LAT. The Phospho-Lat (Y191) Recombinant Antibody has been validated for the following applications: ELISA, WB, IHC, IF, IP. Host: HEK293F Cell. Species: Human (Homo sapiens). Immunogen: A Synthesized Peptide Derived From Human Phospho-Lat (Y191). Applications: ELISA, WB, IHC, IF, IP. Target Synonyms: LAT. Accession Number: O43561. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7691