Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospholipase C beta 3 Antibody (YA2694) | 100 UL
SDP

Catalog No. 5000443962
Encompass_Preferred

The Phospholipase C beta 3 Antibody (YA2694) is a rabbit-derived monoclonal antibody reactive with human samples. This product is supplied as a liquid at a concentration of 1.06 mg/mL and is designed for research use only.

  • Predicted molecular weight: 139 kDa; observed: 150 kDa
  • Formulated in phosphate buffered saline, pH 7.4, 150 mM NaCl, 0.02% sodium azide and 50% glycerol
  • Applications include Western Blot (WB) and Immunohistochemistry-Paraffin (IHC-P)
  • Recommended storage at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Shipped with blue ice

Catalog No. 50-004-43962 Supplier Medchemexpress LLC Supplier No. HYP82949100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Phospholipase C beta 3 Antibody (YA2694) is a rabbit-derived monoclonal antibody reactive with human samples. This product is supplied as a liquid at a concentration of 1.06 mg/mL and is designed for research use only.

  • Predicted molecular weight: 139 kDa; observed: 150 kDa
  • Formulated in phosphate buffered saline, pH 7.4, 150 mM NaCl, 0.02% sodium azide and 50% glycerol
  • Applications include Western Blot (WB) and Immunohistochemistry-Paraffin (IHC-P)
  • Recommended storage at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Shipped with blue ice

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.