Learn More
Medchemexpress LLC Phospholipase D, peanut | 9001-87-0 | 200 U
SDP
Catalog No. 5000399860
Encompass_Preferred
Phospholipase D, peanut is an important signaling enzyme in mammalian cells. It catalyzes the hydrolysis of phosphatidylcholine, resulting in the production of phosphatidic acid (PA) and choline. This product is intended for research purposes only and is not for human use.
  • Catalyzes the hydrolysis of phosphatidylcholine
  • Produces phosphatidic acid (PA) and choline
  • Solid appearance
  • White to off-white color
  • EC Number 3.1.4.4
  • Enzyme activity of ≥60 U/mg solid
  • Unit defined as the amount of enzyme that catalyzes the release of 1 μmol of choline from L-α-phosphatidylcholine (yolk) per hour at pH 5.6 and 30°C
Catalog No. 50-003-99860 Supplier Medchemexpress LLC Supplier No. HYP2812A200U
Phospholipase D, peanut is an important signaling enzyme in mammalian cells. It catalyzes the hydrolysis of phosphatidylcholine, resulting in the production of phosphatidic acid (PA) and choline. This product is intended for research purposes only and is not for human use.
  • Catalyzes the hydrolysis of phosphatidylcholine
  • Produces phosphatidic acid (PA) and choline
  • Solid appearance
  • White to off-white color
  • EC Number 3.1.4.4
  • Enzyme activity of ≥60 U/mg solid
  • Unit defined as the amount of enzyme that catalyzes the release of 1 μmol of choline from L-α-phosphatidylcholine (yolk) per hour at pH 5.6 and 30°C