Learn More
Fujifilm Healthcare Americas Corp Phospholipids C
Catalog No. NC9993780
Encompass_Preferred
The Phospholipids C kit is an in vitro assay utilizing an enzymatic colorimetric method the quantitative determination of phospholipids (lecithin, lysolecithin, and sphingomyelin) in serum. Package size: Kit includes: Buffer Solution- 8 x 50 mL, Color Reagent- 8 x for 50 mL, Standard Solution- 2 x 10 mL bottles. This product is for Research Use Only, not for use in diagnostic procedures. Contact supplier for additional information at www.ivd.fujimed.com.
Catalog No. NC9993780 Supplier Fujifilm Healthcare Americas Corp Supplier No. 99701801
Add to Cart
Add to Cart
The Phospholipids C kit is an in vitro assay utilizing an enzymatic colorimetric method the quantitative determination of phospholipids (lecithin, lysolecithin, and sphingomyelin) in serum. Package size: Kit includes: Buffer Solution- 8 x 50 mL, Color Reagent- 8 x for 50 mL, Standard Solution- 2 x 10 mL bottles. This product is for Research Use Only, not for use in diagnostic procedures. Contact supplier for additional information at www.ivd.fujimed.com.