Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphomolybdic Acid ACS MP Biomedicals

Catalog No. ICN15255680 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15255680 100 g
ICN15255625 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15255680 Supplier MP Biomedicals, Inc Supplier No. 0215255680
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 51429-74-4
Molecular Formula H3Mo12O40P
Molecular Weight (g/mol) 1825.40
MDL Number MFCD00149913
InChI Key FEFSFHGZSNHJEL-UHFFFAOYSA-N
Synonym Molybdophosphoric acid
IUPAC Name 12-Molybdophosphoric acid
SMILES [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].OP(O)(O)=O

Specifications

Density 0.86 g/mL
Quantity 100 g
Formula Weight 1825.25
Grade ACS
Chemical Name or Material Phosphomolybdic Acid
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.