Learn More
Electron Microscopy Sciences Phosphomolybdic Acid, Crystal, Reagent A.C.S. 100 GR
SDP
Catalog No. 5027854
Encompass_Preferred
(dodeca-Molybdophosphoric Acid)
CAS #51429-74-4 H₃PO₄·12(MoO₃)·24H₂O F.W. 2257.6
Catalog No. 50-278-54 Supplier Electron Microscopy Sciences Supplier No. 19400
(dodeca-Molybdophosphoric Acid)
CAS #51429-74-4 H₃PO₄·12(MoO₃)·24H₂O F.W. 2257.6