Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphomolybdic acid hydrate, ≥99.99% trace metals basis, MilliporeSigma™ Supelco™

Catalog No. 111018359 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8359 10 g
11-101-7964 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8359 Supplier MilliporeSigma Supelco Supplier No. 43140010G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phosphomolybdic acid hydrate is a heteropolyacid hydrate. It participates in the synthesis of molybdenum disulfide (MoS2)-reduced graphene oxide (RGO) composites. Phosphomolybdic acid is one of the components of the Goldner's trichrome stain used for tissue staining.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00149913
Synonym Molybdophosphoric acid

Specifications

Density 1.62 g/mL (at 25°C)
CAS 51429-74-4
Packaging Glass Bottle
Quantity 10 g
Linear Formula H3 [P(Mo3O10)4 ] · xH2 O
Formula Weight 1825.25 (anhydrous basis)
Percent Purity ≥99.99% (trace metals basis)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.