Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphomolybdic Acid Hydrate (Crystalline/Certified ACS), Fisher Chemical

Catalog No. A237100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A237-100 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A237-100
  • $1,942.00
    $820.65 / Each

    Save $1,121.35

    58% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $8,960.00
    $3,905.65 / Case of 6 EA

    Save $5,054.35

    56% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 51429-74-4
Molecular Formula H3Mo12O40P
Molecular Weight (g/mol) 1825.40
MDL Number MFCD00149913
InChI Key FEFSFHGZSNHJEL-UHFFFAOYSA-N
Synonym Molybdophosphoric Acid Hydrate
IUPAC Name 12-Molybdophosphoric acid
SMILES [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].OP(O)(O)=O

Specifications

Color Yellow
Melting Point 78°C to 90°C
Density 3.15g/cm³
Heavy Metals (as Pb) 0.005% max.
Identification Pass Test
Packaging Glass Bottle
Decomposition Information 500°C
Quantity 100 g
Physical Form Solid
UN Number 3085
Formula Weight 1825.25
Grade Certified ACS
Chemical Name or Material Phosphomolybdic Acid Hydrate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Corrosive to metals. Use personal protective equipment. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.

NFPA

Health:3
Flammability:0
Instability:2
DOTInformation Class 5.1: Oxidizer
Recommended Storage Refrigerate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.