Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphomolybdic acid hydrate, For microscopy, MilliporeSigma™ Supelco™

Catalog No. 111018521 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8521 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8521 Supplier MilliporeSigma Supelco Supplier No. 79560100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phosphomolybdic acid hydrate (PAH) is yellow and crystallisable compound. It precipitates from potash, oxides of rubidium, caesium and thallium, ammonia and other nitrogenous organic alkaloids.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00149913
Synonym Molybdophosphoric acid

Specifications

Density 1.62 g/mL (at 25°C)
CAS 51429-74-4
Quantity 100 g
Linear Formula H3 [P(Mo3O10)4 ] · xH2 O
Formula Weight 1825.25 (anhydrous basis)
Grade Microscopy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.