Learn More
Sigma Aldrich Fine Chemicals Biosciences Phosphomolybdic acid solution ready-to-use spray and plunge reagent, for chromatography | 12026-57-2 | MFCD00011341 | 500ML
Catalog No. 501809450
Encompass
Phosphomolybdic acid solution ready-to-use spray and plunge reagent, for chromatography | Mol Wt: 1825.25 | 12026-57-2 | MFCD00011341 | 500ML
Catalog No. 50-180-9450 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 02553500ML
Phosphomolybdic acid solution ready-to-use spray and plunge reagent, for chromatography | Mol Wt: 1825.25 | 12026-57-2 | MFCD00011341 | 500ML