Learn More
Newcomer Supply Phosphomolybdic - Phosphotungstic Acid, Aqueous, 500 ml
SDP
Catalog No. NC2101918
Encompass
Use: Masson Trichrome Stain. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.
This item is not returnable. View return policy
Catalog No. NC2101918 Supplier Newcomer Supply Supplier No. 1332B
Use: Masson Trichrome Stain. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.