Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phosphomolybdic Acid Solution

Catalog No. P4869100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
25 g
Danger

Catalog No. P4869100ML

Supplier: Sigma Aldrich P4869100ML

Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

For the detection of lipids, steroids, lactones, keto acids, hydroxy acids, unsaturated fatty acids, and phenolic compounds.10% Phosphomolybdic acid in ethanol

Specifications

CAS 1778-02-5
Melting Point 149°C to 152°C
Molecular Formula C23H34O3
Linear Formula C23H34O3
MDL Number MFCD00003627
Quantity 25 g
Molecular Weight (g/mol) 358.51 g/mol
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.