Learn More
Research Products International Corp Fosfomycin [Phosphomycin], Disodium Salt, 1 Gram
SDP
Catalog No. 501368271
Encompass_Preferred
Fosfomycin is an antibiotic that targets a broad spectrum of bacteria, including both Gram-positive and Gram-negative strains. It exerts its antibacterial effects by inhibiting cell wall synthesis, disrupting the structural integrity of bacterial cells.  
In laboratory research, Fosfomycin is employed to study bacterial resistance mechanisms, antibiotic susceptibility, and the development of new therapies. It helps researchers understand the complex dynamics between bacteria and antibiotics, advancing our knowledge of microbiology and drug development. 
Catalog No. 50-136-8271 Supplier Research Products International Corp Supplier No. F901001.0
Fosfomycin is an antibiotic that targets a broad spectrum of bacteria, including both Gram-positive and Gram-negative strains. It exerts its antibacterial effects by inhibiting cell wall synthesis, disrupting the structural integrity of bacterial cells.  
In laboratory research, Fosfomycin is employed to study bacterial resistance mechanisms, antibiotic susceptibility, and the development of new therapies. It helps researchers understand the complex dynamics between bacteria and antibiotics, advancing our knowledge of microbiology and drug development. 