Learn More
Medchemexpress LLC Phosphonic acid, P-(2-amino-2,3-dihydro-1H-inden-2-yl)-, hydrochloride (1:1) | 1416354-35-2 | MFCD23135861 | 98.4% | 249.63 g/mol | C9H13ClNO3P | 50 MG
SDP
Catalog No. 5000300878
Encompass_Preferred
2-Aminoindan-2-phosphonic acid hydrochloride is the hydrochloride salt of 2-aminoindan-2-phosphonic acid, a competitive phenylalanine ammonia-lyase (PAL) inhibitor used in biochemical assays and plant metabolism research. It is supplied as a solid of high purity (≈98.4%) with reported formula C9H13ClNO3P and molecular weight 249.63 g/mol; documentation such as a certificate of analysis and safety data sheet is available.
  • High purity solid suitable for biochemical assays and research use.
  • Acts as a competitive phenylalanine ammonia-lyase (PAL) inhibitor.
  • Well characterized: formula C9H13ClNO3P, molecular weight 249.63 g/mol.
  • Available in small-scale quantities for laboratory research.
  • Certificate of analysis and safety data sheet available for quality and handling information.
Catalog No. 50-003-00878 Supplier Medchemexpress LLC Supplier No. HYW01769250MG
2-Aminoindan-2-phosphonic acid hydrochloride is the hydrochloride salt of 2-aminoindan-2-phosphonic acid, a competitive phenylalanine ammonia-lyase (PAL) inhibitor used in biochemical assays and plant metabolism research. It is supplied as a solid of high purity (≈98.4%) with reported formula C9H13ClNO3P and molecular weight 249.63 g/mol; documentation such as a certificate of analysis and safety data sheet is available.
  • High purity solid suitable for biochemical assays and research use.
  • Acts as a competitive phenylalanine ammonia-lyase (PAL) inhibitor.
  • Well characterized: formula C9H13ClNO3P, molecular weight 249.63 g/mol.
  • Available in small-scale quantities for laboratory research.
  • Certificate of analysis and safety data sheet available for quality and handling information.