Learn More
Medchemexpress LLC Phosphonium, [6-(3,8-diamino-6-phenyl-5(6H)-phenanthridinyl)hexyl]triphenyl-, iodide (1:1) | 1003197-00-9 | 95.6% | 759.70 | 250 MG
SDP
Catalog No. 5000451377
Encompass_Preferred
MitoSOX Red is a cell-permeable, live cell fluorescent probe that specifically targets mitochondria. It is oxidized by superoxide within the mitochondria, but not by other Reactive Oxygen Species (ROS) or Reactive Nitrogen Species (RNS) generating systems. The oxidized MitoSOX Red then binds to nucleic acids in the mitochondria/nucleus, producing a strong red fluorescence, making it a fluorescent indicator for specifically detecting superoxide. Superoxide dismutase (SOD) can prevent its oxidation. It is intended for research use only.
  • Cell-permeable live cell fluorescent probe
  • Specifically targets mitochondria
  • Oxidized by superoxide, not by other ROS or RNS
  • Binds to nucleic acids in mitochondria/nucleus
  • Produces strong red fluorescence for superoxide detection
  • Oxidation can be prevented by superoxide dismutase (SOD)
Catalog No. 50-004-51377 Supplier Medchemexpress LLC Supplier No. 250MGHYW011511
MitoSOX Red is a cell-permeable, live cell fluorescent probe that specifically targets mitochondria. It is oxidized by superoxide within the mitochondria, but not by other Reactive Oxygen Species (ROS) or Reactive Nitrogen Species (RNS) generating systems. The oxidized MitoSOX Red then binds to nucleic acids in the mitochondria/nucleus, producing a strong red fluorescence, making it a fluorescent indicator for specifically detecting superoxide. Superoxide dismutase (SOD) can prevent its oxidation. It is intended for research use only.
  • Cell-permeable live cell fluorescent probe
  • Specifically targets mitochondria
  • Oxidized by superoxide, not by other ROS or RNS
  • Binds to nucleic acids in mitochondria/nucleus
  • Produces strong red fluorescence for superoxide detection
  • Oxidation can be prevented by superoxide dismutase (SOD)