Learn More
Selleck Chemical LLC Phosphonoacetic acid S3356-100mg
SDP
Catalog No. 503110589
Encompass_Preferred
Phosphonoacetic acid (PAA) is an active endogenous metabolite that possesses a restricted potential to block DNA biosynthesis Phosphonoacetic acid exhibits anti-viral activities
Catalog No. 50-311-0589 Supplier Selleck Chemical LLC Supplier No. S3356100MG
Phosphonoacetic acid (PAA) is an active endogenous metabolite that possesses a restricted potential to block DNA biosynthesis Phosphonoacetic acid exhibits anti-viral activities