Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphonomycin disodium salt, 100.4%, MP Biomedicals™

Catalog No. ICN15187605 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15187605 5 g
ICN15187610 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15187605 Supplier MP Biomedicals, Inc Supplier No. 0215187605
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 26016-99-9 | Formula: C3H5Na2O4P | Weight: 182.02 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26016-99-9
Molecular Formula C3H5Na2O4P
Molecular Weight (g/mol) 182.02
MDL Number MFCD00056853
InChI Key QZIQJIKUVJMTDG-JSTPYPERSA-L
Synonym (2R-cis)-(3-Methyloxiranyl) phosphonic acid disodium salt, Fosfomycin disodium
PubChem CID 16397688
IUPAC Name disodium [(2R,3S)-3-methyloxiran-2-yl]phosphonate
SMILES [Na+].[Na+].C[C@@H]1O[C@@H]1P([O-])([O-])=O

Specifications

Type Phosphonomycin Disodium Salt
Physical Form Powder
Color White
Quantity 5 g
Assay Percent Range 1.004
Formula Weight 182
Grade Not Available
Chemical Name or Material Phosphonomycin disodium salt
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.