Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology PhosphoPlus™ Myosin Light Chain 2 Antibody Duet, Unconjugated, 1 Kit
SDP

Catalog No. 502726106
Encompass

The PhosphoPlus™ Myosin Light Chain 2 (Thr18/Ser19) Antibody Duet provides an efficient method to assess the activation status of myosin light chain 2 (MLC2). This kit includes one antibody specific to the phosphorylation site at Thr18 and Ser19, and a second antibody for total MLC2 protein detection. Each antibody is selected for superior performance in western blotting and immunofluorescence applications.

  • Contains an activation-state and total protein antibody
  • Validated for western blotting and immunofluorescence
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and < 0.02% sodium azide
  • Store at -20°C
  • Do not aliquot the antibodies

Catalog No. 50-272-6106 Supplier Cell Signaling Technology Supplier No. 45893S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The PhosphoPlus™ Myosin Light Chain 2 (Thr18/Ser19) Antibody Duet provides an efficient method to assess the activation status of myosin light chain 2 (MLC2). This kit includes one antibody specific to the phosphorylation site at Thr18 and Ser19, and a second antibody for total MLC2 protein detection. Each antibody is selected for superior performance in western blotting and immunofluorescence applications.

  • Contains an activation-state and total protein antibody
  • Validated for western blotting and immunofluorescence
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and < 0.02% sodium azide
  • Store at -20°C
  • Do not aliquot the antibodies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.