Learn More
Cell Signaling Technology PhosphoPlus™ Rad50 Antibody Duet, Rabbit IgG, Unconjugated, 1 Kit
SDP
Catalog No. 503134389
Encompass
The PhosphoPlus™ Rad50 (Ser635) Antibody Duet is designed to facilitate the assessment of protein activation status. Each duet contains an activation-state specific antibody and a total protein antibody for the target of interest. These antibodies are selected for their performance in specified applications to provide reliable detection of Rad50 and its phosphorylated form.
  • Includes Phospho-Rad50 (Ser635) (F5H8B) rabbit monoclonal antibody (100 µl)
  • Includes Rad50 (E3I8K) rabbit monoclonal antibody (100 µl)
  • Suitable for western blotting, flow cytometry, and immunoprecipitation
  • Targets Rad50 and Rad50 (Ser635) phosphate
  • Supplied in storage buffer with 50% glycerol
Catalog No. 50-313-4389 Supplier Cell Signaling Technology Supplier No. 20286S
The PhosphoPlus™ Rad50 (Ser635) Antibody Duet is designed to facilitate the assessment of protein activation status. Each duet contains an activation-state specific antibody and a total protein antibody for the target of interest. These antibodies are selected for their performance in specified applications to provide reliable detection of Rad50 and its phosphorylated form.
  • Includes Phospho-Rad50 (Ser635) (F5H8B) rabbit monoclonal antibody (100 µl)
  • Includes Rad50 (E3I8K) rabbit monoclonal antibody (100 µl)
  • Suitable for western blotting, flow cytometry, and immunoprecipitation
  • Targets Rad50 and Rad50 (Ser635) phosphate
  • Supplied in storage buffer with 50% glycerol