Learn More
ABP BIOSCIENCES Phospho-Tag™ Phosphoprotein Gel Stain
Catalog No. NC1604470
Encompass_Preferred
Phospho-Tag™ Phosphoprotein Gel Stain is a high sensitive fluorescent stain designed for selectively detecting phosphoproteins in polyacrylamide gels. This stain contains a phospho-tag™ group, which allows direct, in-gel detection of phosphate groups attached to tyrosine, serine, or threonine residues, without the need for antibodies or radioisotopes. The stain can be used with standard SDS-polyacrylamide gels or with 2-D gels.
Catalog No. NC1604470 Supplier ABP BIOSCIENCES Supplier No. P005A
Phospho-Tag™ Phosphoprotein Gel Stain is a high sensitive fluorescent stain designed for selectively detecting phosphoproteins in polyacrylamide gels. This stain contains a phospho-tag™ group, which allows direct, in-gel detection of phosphate groups attached to tyrosine, serine, or threonine residues, without the need for antibodies or radioisotopes. The stain can be used with standard SDS-polyacrylamide gels or with 2-D gels.