Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phosphoramidate proagent of 6-O-methyl-2'-C-methyl guanosine | 1234490-83-5 | MFCD22124897 | 99.9% | 658.64 g/mol | C30H39N6O9P | 5 MG
SDP

Catalog No. 5000262489
Encompass_Preferred

A phosphoramidate prodrug of 6-O-methyl-2′-C-methyl guanosine that potently inhibits hepatitis C virus (HCV) replication. Supplied as a white to off-white powder for research use, it offers high analytical purity and defined storage recommendations for both powder and solution formats.

  • High purity (≈99.9%).
  • Molecular formula C30H39N6O9P and molecular weight 658.64 g/mol.
  • Solid, white to off-white powder suitable for in vitro studies.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available in small research pack sizes including 5 mg.

Catalog No. 50-002-62489 Supplier Medchemexpress LLC Supplier No. HY133375MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

A phosphoramidate prodrug of 6-O-methyl-2′-C-methyl guanosine that potently inhibits hepatitis C virus (HCV) replication. Supplied as a white to off-white powder for research use, it offers high analytical purity and defined storage recommendations for both powder and solution formats.

  • High purity (≈99.9%).
  • Molecular formula C30H39N6O9P and molecular weight 658.64 g/mol.
  • Solid, white to off-white powder suitable for in vitro studies.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available in small research pack sizes including 5 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.