Learn More
Medchemexpress LLC HY-137316A 5mg Medchemexpress, Phosphoramide mustard (cyclohexanamine) CAS:1566-15-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY137316A5MG
Encompass_Preferred
Medchemexpress, HY-137316A 5mg Phosphoramide mustard (cyclohexanamine) CAS:1566-15-0 Phosphoramide mustard cyclohexanamine is a biologically active metabolite of Cyclophosphamide (HY-17420), with anticancer activitiy. Phosphoramide mustard cyclohexanamine induces DNA damage. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1485