Learn More
Research Products International Corp Phosphoramidon, 5 Milligrams
SDP
Catalog No. 501368490
Encompass_Preferred
Protease inhibitor specific for thermolysin and other metalloproteases. Also effective inhibitor of collegenase.
Working Concentration: 5-500 um
* Please Note: This product ships on ice to ensure chemical stability. Product will not ship on a Friday or on any day before a Holiday that would cause the product to sit in storage during transit and become unstable.
Catalog No. 50-136-8490 Supplier Research Products International Corp Supplier No. P202750.005
Add to Cart
Add to Cart
Protease inhibitor specific for thermolysin and other metalloproteases. Also effective inhibitor of collegenase.
Working Concentration: 5-500 um
* Please Note: This product ships on ice to ensure chemical stability. Product will not ship on a Friday or on any day before a Holiday that would cause the product to sit in storage during transit and become unstable.