Learn More
Medchemexpress LLC HY-N2021A 5mg Medchemexpress, Phosphoramidon (Disodium) CAS:164204-38-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN2021A5MG
Encompass_Preferred
Medchemexpress, HY-N2021A 5mg Phosphoramidon (Disodium) CAS:164204-38-0 Phosphoramidon Disodium is a metalloprotease inhibitor. Phosphoramidon inhibits endothelin-converting enzyme (ECE), neutral endopeptidase (NEP), and angiotensin-converting enzyme (ACE) with IC50 values of 3.5, 0.034, and 78 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0358