Learn More
Sigma Aldrich Fine Chemicals Biosciences Phosphoramidon disodium sa25MG
Catalog No. 502908157
Encompass
Phosphoramidon [N-(rhamnopyranosyl-oxyhydroxyphosphinyl)-Leu-Trp] is a competitive inhibitor of several soluble zinc metalloproteases. Phosphoramidate is produced naturally by Streptomycesnbsptanashiensis.
Catalog No. 50-290-8157 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. R738525MG
Phosphoramidon [N-(rhamnopyranosyl-oxyhydroxyphosphinyl)-Leu-Trp] is a competitive inhibitor of several soluble zinc metalloproteases. Phosphoramidate is produced naturally by Streptomycesnbsptanashiensis.