Learn More
Selleck Chemical LLC Phosphoramidon Disodium Salt
Supplier: Selleck Chemical LLC S73825MG
Phosphoramidon Disodium Salt is a metalloendopeptidase inhibitor widely used as a biochemical tool Phosphoramidon inhibited ECE NEP and ACE activities with IC50 values of 3 5 M 0 034 M and 78 M respectively