Learn More
Selleck Chemical LLC Phosphoramidon Disodium Salt
SDP
Catalog No. 5000004423
Encompass_Preferred
Phosphoramidon Disodium Salt is a metalloendopeptidase inhibitor widely used as a biochemical tool Phosphoramidon inhibited ECE NEP and ACE activities with IC50 values of 3 5 M 0 034 M and 78 M respectively
Catalog No. 50-000-04423 Supplier Selleck Chemical LLC Supplier No. S738210MM/1ML
Phosphoramidon Disodium Salt is a metalloendopeptidase inhibitor widely used as a biochemical tool Phosphoramidon inhibited ECE NEP and ACE activities with IC50 values of 3 5 M 0 034 M and 78 M respectively