Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoramidon disodium salt, 97+%, Thermo Scientific Chemicals

Catalog No. AAJ63456LB0
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63456LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ63456LB0 Supplier Thermo Scientific Chemicals Supplier No. J63456LB0
Add to Cart

missing translation for 'validMainframeMsg'

 

Inhibitor of thermolysin and collagenase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119942-99-3
Molecular Formula C23H34N3NaO10P
Molecular Weight (g/mol) 566.50
MDL Number MFCD00077870
InChI Key KITONROGOABYFZ-UHFFFAOYNA-N
Synonym phosphoramidon disodium salt dihydrate
PubChem CID 129893979
IUPAC Name (2S)-2-[[(2S)-2-[[hydroxy-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyphosphoryl]amino]-4-methylpentanoyl]amino]-3-(1H-indol-3-yl)propanoic acid;sodium;dihydrate
SMILES [Na].CC(C)CC(NP(O)(=O)OC1OC(C)C(O)C(O)C1O)C(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

Color White to Yellow
Physical Form Solid
Quantity 5 mg
Beilstein 8105669
Formula Weight 587.47
Percent Purity ≥97%
Grade Ungraded
Chemical Name or Material Phosphoramidon disodium salt
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.